Assay Details
Gene Symbol: Larp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040399.1 | NP_001035489.1 | 4 - 5 | 345 | 65 | |
GenBank mRNA | AK076732.1 | - | 4 - 5 | 313 | 65 | |
AK160155.1 | - | 4 - 5 | 345 | 65 | ||
BC025528.1 | - | 2 - 3 | 268 | 5324328 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
