Assay Details
| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145991.1 | NP_666103.1 | 7 - 8 | 788 | 103 | |
| GenBank mRNA | AK080861.1 | - | 7 - 8 | 920 | 103 | |
| BC027756.1 | - | 7 - 8 | 889 | 5067496 | 103 | |
| BC031127.1 | - | 7 - 8 | 788 | 4972826 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc73
Genomic Map