Assay Details
Gene Symbol: Rhot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145999.2 | NP_666111.1 | 14 - 15 | 1611 | 72 | |
GenBank mRNA | AY311390.1 | - | 14 - 15 | 1611 | 72 | |
BC023163.1 | - | 4 - 5 | 381 | 72 | ||
BC023927.1 | - | 2 - 3 | 415 | 72 | ||
BC029777.1 | - | 14 - 15 | 1244 | 4950061 | 72 | |
BC034062.1 | - | 14 - 15 | 1266 | 4207654 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
