Assay Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146017.3 | NP_666129.1 | 5 - 6 | 651 | 102 | |
| GenBank mRNA | AK028946.1 | - | 5 - 6 | 651 | 102 | |
| AK160577.1 | - | 5 - 6 | 655 | 102 | ||
| BC023693.1 | - | 5 - 6 | 638 | 5344479 | 102 | |
| BC025550.1 | - | 5 - 6 | 596 | 5320737 | 102 | |
| BC027245.1 | - | 5 - 6 | 614 | 3602400 | 102 | |
| BC027839.1 | - | 1 - 2 | 203 | 102 | ||
| BC031196.1 | - | 5 - 6 | 625 | 5059564 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrp
Genomic Map