Assay Details
| Gene Symbol: Srp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146032.3 | NP_666144.3 | 9 - 10 | 1192 | 74 | |
| GenBank mRNA | AK033010.1 | - | 9 - 10 | 1111 | 74 | |
| AK076003.1 | - | 9 - 10 | 1089 | 74 | ||
| AK187819.1 | - | 1 - 2 | 119 | 74 | ||
| BC033573.1 | - | 5 - 6 | 492 | 5323201 | 74 | |
| BC070422.1 | - | 9 - 10 | 1192 | 6403747 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srp68
Genomic Map