Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200002386.mm.1 |
NM_146033.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002386.mm.1 |
NM_146033.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002386.mm.1 |
TC1200000319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002388.mm.1 |
NM_146033.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002388.mm.1 |
NM_146033.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002388.mm.1 |
TC1200000319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002389.mm.1 |
NM_146033.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002389.mm.1 |
NM_146033.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002389.mm.1 |
TC1200000319.mm.1 |
MTA TC |