Assay Details
| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146040.1 | NP_666152.1 | 6 - 7 | 962 | 135 | |
| GenBank mRNA | AK149673.1 | - | 6 - 7 | 940 | 135 | |
| BC006933.1 | - | 6 - 7 | 962 | 3602150 | 135 | |
| BC071198.1 | - | 6 - 7 | 930 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdca7l
Genomic Map