Assay Details
| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146040.1 | NP_666152.1 | 7 - 8 | 1082 | 70 | |
| GenBank mRNA | AK149673.1 | - | 7 - 8 | 1060 | 70 | |
| BC006933.1 | - | 7 - 8 | 1082 | 3602150 | 70 | |
More Information
Set Membership: |
|

Cdca7l
Genomic Map