Assay Details
Gene Symbol: Tfb1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146074.1 | NP_666186.1 | 1 - 2 | 132 | 82 | |
GenBank mRNA | AF508971.1 | - | 1 - 2 | 132 | 82 | |
BC032930.1 | - | 1 - 2 | 132 | 1382837 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
