Assay Details
| Gene Symbol: Apbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146085.1 | NP_666197.1 | 1 - 2 | 374 | 104 | |
| GenBank mRNA | AK141878.1 | - | 1 - 2 | 276 | 104 | |
| AK171634.1 | - | 1 - 2 | 325 | 104 | ||
| BC024136.1 | - | 1 - 2 | 130 | 104 | ||
| BC024809.1 | - | 1 - 2 | 374 | 5357681 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apbb3
Genomic Map