Assay Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146089.2 | NP_666201.1 | 6 - 7 | 694 | 122 | |
| GenBank mRNA | AK076912.1 | - | 6 - 7 | 674 | 122 | |
| AK146923.1 | - | 6 - 7 | 691 | 122 | ||
| BC024400.1 | - | 6 - 7 | 694 | 5026575 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Haus1
Genomic Map