Assay Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043354.2 | NP_001036819.1 | 7 - 8 | 1637 | 119 | |
| NM_001289921.1 | NP_001276850.1 | 8 - 9 | 1156 | 119 | ||
| NM_146095.4 | NP_666207.3 | 7 - 8 | 1103 | 119 | ||
| GenBank mRNA | AB258405.1 | - | 7 - 8 | 1264 | 119 | |
| AK134790.1 | - | 7 - 8 | 1404 | 119 | ||
| BC024842.1 | - | 8 - 9 | 1156 | 5358124 | 119 | |
| BC058269.1 | - | 6 - 7 | 1010 | 119 | ||
| DQ779923.1 | - | 6 - 7 | 1007 | 119 | ||
| DQ779924.1 | - | 7 - 8 | 1032 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rorb
Genomic Map