Assay Details
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 25 - 26 | 2885 | 78 | |
GenBank mRNA | AK031548.1 | - | 25 - 26 | 2885 | 78 | |
AK050944.1 | - | 11 - 12 | 1068 | 78 | ||
AK166470.1 | - | 23 - 24 | 2884 | 78 | ||
BC023786.1 | - | 24 - 25 | 2263 | 78 | ||
BC024544.1 | - | 9 - 10 | 870 | 78 | ||
BC026832.1 | - | 25 - 26 | 2935 | 4486176 | 78 | |
BC029977.1 | - | 16 - 17 | 1521 | 78 | ||
BC036128.1 | - | 25 - 26 | 3003 | 4971791 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
