Assay Details
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146151.4 | NP_666263.3 | 3 - 4 | 721 | 94 | |
GenBank mRNA | AK031814.1 | - | 3 - 4 | 714 | 94 | |
AK047755.1 | - | 3 - 4 | 722 | 94 | ||
AK053368.1 | - | 790 | 94 | |||
BC019149.1 | - | 2 - 3 | 454 | 5053345 | 94 | |
BC029766.1 | - | 3 - 4 | 699 | 4947564 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
