Assay Details
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026145.4 | NP_080421.2 | 3 - 4 | 460 | 62 | |
GenBank mRNA | AK028657.1 | - | 3 - 4 | 427 | 62 | |
AK041516.1 | - | 3 - 4 | 427 | 62 | ||
AK145295.1 | - | 3 - 4 | 439 | 62 | ||
AK147714.1 | - | 3 - 4 | 405 | 62 | ||
AK161199.1 | - | 3 - 4 | 417 | 62 | ||
AK163351.1 | - | 3 - 4 | 414 | 62 | ||
AK166505.1 | - | 3 - 4 | 439 | 62 | ||
BC006935.1 | - | 3 - 4 | 441 | 3601917 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
