Assay Details
| Gene Symbol: Lrit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146245.2 | NP_666357.2 | 2 - 3 | 684 | 78 | |
| GenBank mRNA | AK044631.1 | - | 2 - 3 | 685 | 78 | |
| BC032270.1 | - | 2 - 3 | 690 | 5401567 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrit1
Genomic Map