Assay Details
| Gene Symbol: Stard13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163493.1 | NP_001156965.1 | 7 - 8 | 2244 | 71 | |
| NM_146258.2 | NP_666370.3 | 7 - 8 | 2187 | 71 | ||
| GenBank mRNA | AK050259.1 | - | 7 - 8 | 2245 | 71 | |
| AK136202.1 | - | 3 - 4 | 770 | 71 | ||
| BC027830.1 | - | 7 - 8 | 2199 | 5320966 | 71 | |
| DV651942.1 | - | 3 - 4 | 446 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard13
Genomic Map