Assay Details
Gene Symbol: Olfr846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146282.2 | NP_666394.1 | 1 | 904 | 105 | |
GenBank mRNA | BC134375.1 | - | 1 | 905 | 40135046 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969640 | NM_146282.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969640 | BC134375.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069689 | NM_146282.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069689 | BC134375.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129146 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069689 | NM_146282.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069689 | BC134375.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969640 | NM_146282.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969640 | BC134375.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269356 | NM_146282.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269356 | BC134375.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339783 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339795 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339798 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26339793 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129146 | - | - |
Whole Mouse Genome G4122A | A_51_P269356 | NM_146282.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P269356 | BC134375.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102512 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116722 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129146 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07743761 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07743771 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07743750 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07743755 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07743776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26339807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07618173 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339762 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26339755 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10591191 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10591190 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 599688 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6760097 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6760097 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006760097 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229446 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1229446 | - | - |
mouse-6 v1.1 (Illumina) | scl36179.1.1_184-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6760097 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl36179.1.1_184-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760097 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1229446 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229446 | - | - |
