Assay Details
Gene Symbol: Olfr981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146286.1 | NP_666398.1 | 1 | 46 | 87 | |
GenBank mRNA | BC120776.1 | - | 1 | 89 | 40129699 | 87 |
BC120778.1 | - | 1 | 89 | 40129701 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P225573 | BC120778.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P225573 | BC120778.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153313 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P225573 | BC120778.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P225573 | BC120776.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P225573 | BC120778.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225573 | BC120778.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225573 | BC120778.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441021 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441006 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153313 | - | - |
Whole Mouse Genome G4122A | A_51_P225573 | BC120778.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225573 | BC120778.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225573 | NM_146286.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225573 | BC120776.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112926 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153313 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07915161 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07915163 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07885535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07665797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440993 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441031 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07885525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07677269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07915161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440994 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10584523 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10584522 | - | - |
Affymetrix Mouse Clariom S Array | PSR0900002922.mm.1 | TC0900000445.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900002923.mm.1 | NM_146286.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900002923.mm.1 | NM_146286.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900002923.mm.1 | TC0900000445.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 789454 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6550435 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6550435 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006550435 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217634 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217634 | - | - |
mouse-6 v1.1 (Illumina) | scl7554.1.1_275-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6550435 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl7554.1.1_275-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550435 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217634 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217634 | - | - |
