Assay Details
Gene Symbol: Olfr111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005485.2 | NP_001005485.1 | 1 | 369 | 82 | |
GenBank mRNA | BC139116.1 | - | 1 | 369 | 8862138 | 82 |
BC139117.1 | - | 1 | 369 | 8862139 | 82 |
Gene Symbol: Olfr110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146328.2 | NP_666440.2 | 2 | 431 | 82 | |
GenBank mRNA | BC132347.1 | - | 1 | 375 | 40130624 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163368 | NM_001005485.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163368 | BC139116.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163368 | BC139117.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987379 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208681 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821120 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377250 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105554 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987379 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163368 | NM_001005485.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163368 | BC139116.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163368 | BC139117.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345011 | NM_001005485.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345011 | BC139116.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345011 | BC139117.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345011 | BC132347.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345011 | NM_146328.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168939 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168950 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168934 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168944 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168932 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168935 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168952 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168971 | - | - |
Whole Mouse Genome G4122A | A_51_P345011 | NM_001005485.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P345011 | BC139116.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P345011 | BC139117.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P345011 | BC132347.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P345011 | NM_146328.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111631 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118136 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13487456 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13557366 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13159643 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13487458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169013 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13311233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13367744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169010 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13297074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13487466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168923 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13058191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13343804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169075 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168953 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13118877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13205058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13367737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169071 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169028 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169074 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168931 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168920 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13367746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13367730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168922 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13118873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169021 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168916 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168909 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13591164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168930 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169065 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168928 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13487463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168950 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168947 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169026 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10445140 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445142 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10445139 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10445141 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700007721.mm.1 | NM_146328.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007721.mm.1 | NM_146328.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007721.mm.1 | TC1700000775.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700007726.mm.1 | NM_001005485.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007726.mm.1 | NM_001005485.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007726.mm.1 | TC1700000777.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700007722.mm.1 | NM_146328.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007722.mm.1 | NM_146328.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007722.mm.1 | TC1700000775.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700007723.mm.1 | NM_146328.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007723.mm.1 | NM_146328.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007723.mm.1 | TC1700000775.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1990253 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001990253 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250690 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250690 | - | - |
mouse-6 v1.1 (Illumina) | scl0258325.1_256-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1990253 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258325.1_256-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990253 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1250690 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2967322 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2967322 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250690 | - | - |
