Assay Details
| Gene Symbol: Olfr1450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146371.1 | NP_666483.1 | 1 | 444 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953152 | NM_146371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127002 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953152 | NM_146371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963206 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127002 | - | - |
| Whole Mouse Genome G4122A | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205850 | NM_146371.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102401 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116052 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127002 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14258212 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14258176 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14258194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14258209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14258172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14366403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14496281 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461787 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461786 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900003024.mm.1 | NM_146371.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003024.mm.1 | NM_146371.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003024.mm.1 | TC1900000261.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 652688 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7050142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7050142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239694 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239694 | - | - |
| mouse-6 v1.1 (Illumina) | scl6292.1.1_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl6292.1.1_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2852196 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2852196 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239694 | - | - |

Olfr1450
Genomic Map