Assay Details
| Gene Symbol: Olfr414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146761.2 | NP_666972.2 | 1 | 553 | 78 | |
| GenBank mRNA | BC106815.1 | - | 1 | 553 | 40043364 | 78 |
| BC106816.1 | - | 1 | 553 | 40043365 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Olfr414
Genomic Map