Assay Details
| Gene Symbol: Olfr1099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146768.1 | NP_666979.1 | 1 | 329 | 81 | |
| GenBank mRNA | BC132348.1 | - | 1 | 351 | 40130625 | 81 |
| BC132350.1 | - | 1 | 351 | 40130627 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053379 | BC132350.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053379 | NM_146768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053379 | BC132348.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362517 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177245 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053379 | BC132350.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053379 | NM_146768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053379 | BC132348.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116896 | BC132350.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116896 | NM_146768.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116896 | BC132348.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177245 | - | - |
| Whole Mouse Genome G4122A | A_51_P116896 | BC132350.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116896 | NM_146768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116896 | BC132348.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102004 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129860 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01662767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01308863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404221 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484641 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484640 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200033824.mm.1 | NM_146768.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033824.mm.1 | NM_146768.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033824.mm.1 | TC0200003962.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 345350 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5360441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360441 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2640625 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2640625 | - | - |
| mouse-6 v1.1 (Illumina) | scl4325.1.1_193-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360441 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4325.1.1_193-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2640625 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2640625 | - | - |

Olfr1099
Genomic Map