Assay Details
| Gene Symbol: Olfr1230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146789.1 | NP_667000.1 | 1 | 153 | 98 | |
| GenBank mRNA | BC108937.1 | - | 1 | 153 | 40040283 | 98 |
| BC108938.1 | - | 1 | 153 | 40040284 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Olfr1230
Genomic Map