Assay Details
| Gene Symbol: Olfr878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146798.2 | NP_667009.1 | 1 | 744 | 107 | |
| GenBank mRNA | BC119219.1 | - | 1 | 742 | 8733968 | 107 |
| BC119221.1 | - | 1 | 744 | 8733970 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26432980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433015 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01908059 | - | - |
| Whole Mouse Genome G4122A | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314991 | BC119219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314991 | BC119221.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314991 | NM_146798.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104614 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01908059 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07788149 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07788128 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07788124 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07788152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07788151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07788136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07788124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08055005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08055004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07712755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07632134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07813713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26432983 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584363 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 342582 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3800717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800717 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721736 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721736 | - | - |
| mouse-6 v1.1 (Illumina) | scl7580.1.1_112-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800717 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl7580.1.1_112-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800717 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2721736 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721736 | - | - |

Olfr878
Genomic Map