Assay Details
| Gene Symbol: Olfr640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146822.2 | NP_667033.2 | 1 | 903 | 72 | |
| GenBank mRNA | BC106802.2 | - | 1 | 903 | 40042505 | 72 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046223 | NM_146822.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046223 | BC106802.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657508 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046223 | NM_146822.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046223 | BC106802.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101022 | NM_146822.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101022 | BC106802.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25386246 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06308159 | - | - |
| Whole Mouse Genome G4122A | A_51_P101022 | NM_146822.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P101022 | BC106802.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112222 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06308159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386254 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566295 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566294 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 826367 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6100427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006100427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2644084 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2644084 | - | - |
| mouse-6 v1.1 (Illumina) | scl8864.1.1_123-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6100427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl8864.1.1_123-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2644084 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2644084 | - | - |

Olfr640
Genomic Map