Assay Details
| Gene Symbol: Olfr1341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146853.2 | NP_667064.2 | 1 | 51 | 77 | |
| GenBank mRNA | BC130239.1 | - | 1 | 55 | 77 | |
| BC152333.1 | - | 1 | 55 | 77 | ||
| BC152353.1 | - | 1 | 55 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058225 | NM_146853.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058225 | BC130239.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175332 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058225 | NM_146853.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058225 | BC130239.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289380 | NM_146853.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P289380 | BC130239.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23241840 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175332 | - | - |
| Whole Mouse Genome G4122A | A_51_P289380 | NM_146853.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P289380 | BC130239.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137670 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04032539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03948416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04032525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03948424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03905555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23241846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03997729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04082539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03239565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03948424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03544147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03483950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03579178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03621303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04082545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03948423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03547523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03483949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03476612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03483957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03905555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04082541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03876660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03461402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03483943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03483952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03483955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03948420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23241883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03406698 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507571 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507569 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400009982.mm.1 | NM_146853.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009982.mm.1 | NM_146853.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009982.mm.1 | TC0400001314.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 473500 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2900390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2679993 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2679993 | - | - |
| mouse-6 v1.1 (Illumina) | scl25037.1.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25037.1.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2679993 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2679993 | - | - |

Olfr1341
Genomic Map