Assay Details
| Gene Symbol: Olfr768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146864.1 | NP_667075.1 | 1 | 677 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974927 | NM_146864.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127990 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974927 | NM_146864.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P456034 | NM_146864.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27544709 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127990 | - | - |
| Whole Mouse Genome G4122A | A_51_P456034 | NM_146864.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109578 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08775805 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08500001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08499996 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08775803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27544732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08779870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08499996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27544673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373613 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373612 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023941.mm.1 | NM_146864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023941.mm.1 | NM_146864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023941.mm.1 | TC1000003174.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 522438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3290021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2672332 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2672332 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258863.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258863.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290021 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2672332 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2672332 | - | - |

Olfr768
Genomic Map