Assay Details
| Gene Symbol: Olfr1221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146902.2 | NP_667113.2 | 1 | 442 | 96 | |
More Information
Updated Mapping Information
Set Membership: | 
		   
					
						
							 | 
				
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							ABI
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358700 | - | - | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166272 | - | - | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358700 | - | - | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358700 | - | - | 
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166272 | - | - | 
| Whole Mouse Genome G4122A | A_51_P358700 | - | - | 
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122004 | - | - | 
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166272 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412575 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412617 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412563 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412547 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412588 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412616 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412505 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412545 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412565 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412608 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412584 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412583 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412605 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412627 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412593 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412511 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412580 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412557 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412581 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412503 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412577 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412555 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412543 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412597 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412620 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412595 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412552 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412513 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412509 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412562 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412572 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412561 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412614 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412573 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412585 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412586 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412550 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412558 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412625 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412602 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412623 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412587 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412599 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412591 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412625 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412561 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412591 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412567 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412585 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412565 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412620 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412562 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412572 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412544 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412619 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412611 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412588 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412612 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412558 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412595 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412617 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412577 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412593 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412610 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412587 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412550 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412584 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412557 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412575 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412608 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02171274 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412602 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412614 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412627 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412581 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412597 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412547 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412623 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412552 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412600 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412586 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412580 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412563 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412599 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412543 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412601 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412605 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412618 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412578 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412569 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412616 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412551 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412545 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412555 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21412583 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484790 | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10484789 | - | - | 
| Affymetrix Mouse Clariom S Array | PSR0200034007.mm.1 | NM_146902.2.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0200034007.mm.1 | NM_146902.2 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0200034007.mm.1 | TC0200004042.mm.1 | MTA TC | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 570903 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105270593 | - | - | 
| Illumina mouse-6 v1.0 expression beadchip | 105270593 | - | - | 
| Illumina mouse-6 v1.0 expression beadchip | 3520050 | - | - | 
| Illumina mouseRef-8 v1.0 expression beadchip | 0003520050 | - | - | 
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222921 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222072 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222921 | - | - | 
| mouse-6 v1.1 (Illumina) | GI_22129082-S | - | - | 
| mouse-6 v1.1 (Illumina) | scl4292.1.1_191-S | - | - | 
| Illumina Sentrix mouse-6.v1 bead array | 3520050 | - | - | 
| Illumina Sentrix mouse-6.v1 bead array | 105270593 | - | - | 
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4292.1.1_191-S | - | - | 
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105270593 | - | - | 
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520050 | - | - | 
| MouseRef-8 v2 Expression BeadChip | ILMN_1222921 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222921 | - | - | 

Olfr1221
Genomic Map