Assay Details
Gene Symbol: Olfr1239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146970.1 | NP_667181.1 | 1 | 454 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110628 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110628 | - | - |
Whole Mouse Genome G4122A | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P276508 | NM_146970.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132974 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21413799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01332226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413809 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01251198 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02114155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413802 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01460408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01331880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413790 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21413769 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484821 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484820 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034053.mm.1 | NM_146970.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034053.mm.1 | NM_146970.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034053.mm.1 | TC0200004058.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 568717 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3140369 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3140369 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003140369 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255055 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255055 | - | - |
mouse-6 v1.1 (Illumina) | scl4278.1.1_212-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3140369 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4278.1.1_212-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3140369 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2969402 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2969402 | - | - |
