Assay Details
Gene Symbol: Olfr659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147050.1 | NP_667261.1 | 1 | 575 | 118 | |
GenBank mRNA | BC148221.1 | - | 1 | 579 | 40136035 | 118 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011210 | NM_147050.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011210 | BC148221.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106244 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011210 | NM_147050.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011210 | BC148221.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235764 | NM_147050.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235764 | BC148221.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106244 | - | - |
Whole Mouse Genome G4122A | A_51_P235764 | NM_147050.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P235764 | BC148221.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129482 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06387735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06015600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06473646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06473651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06473641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389070 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10555884 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10555883 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 378999 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3450446 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3450446 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003450446 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231113 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231113 | - | - |
mouse-6 v1.1 (Illumina) | scl32252.1.1_8-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3450446 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32252.1.1_8-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450446 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1231113 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231113 | - | - |
