Assay Details
| Gene Symbol: Olfr558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147093.3 | NP_667304.1 | 2 | 1040 | 82 | |
| GenBank mRNA | AK141545.1 | - | 2 | 1040 | 82 | |
| AY834217.1 | - | 1 | 911 | 82 | ||
| BC106820.2 | - | 1 | 906 | 82 | ||
| BC130658.1 | - | 1 | 911 | 40043445 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169039 | AK141545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169039 | NM_147093.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110704 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159351 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169039 | AK141545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169039 | NM_147093.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506426 | AK141545.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506426 | AY834217.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506426 | BC106820.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506426 | BC130658.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506426 | NM_147093.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380166 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06305592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159351 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06305583 | - | - |
| Whole Mouse Genome G4122A | A_51_P506426 | AK141545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506426 | AY834217.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506426 | BC106820.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506426 | BC130658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506426 | NM_147093.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134063 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06122438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06391145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06391144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06316588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06391151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06391128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05963727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06316589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06198565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06316584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06391157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06572856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10555739 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1446400_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555736 | - | - |
| Mouse Genome 430 2.0 Array | 1446400_at | - | - |
| Mouse Genome 430 2.0 Array | 1446400_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 259097_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 259097_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012695.mm.1 | TC0700001466.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012695.mm.1 | NM_147093.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012695.mm.1 | NM_147093.3 | Refseq |
| Mouse Expression Set 430 | 1446400_PM_at | - | - |
| Mouse Expression Set 430 | 1446400_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446400_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446400_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 717207 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4050301 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4050301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050301 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631693 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631693 | - | - |
| mouse-6 v1.1 (Illumina) | scl32273.3.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050301 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32273.3.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631693 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631693 | - | - |

Olfr558
Genomic Map