Assay Details
| Gene Symbol: Olfr586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147111.1 | NP_667322.1 | 1 | 849 | 78 | |
| GenBank mRNA | BC104726.1 | - | 1 | 848 | 40043611 | 78 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P107807 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016174 | NM_147111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016174 | BC104726.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127314 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016174 | NM_147111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016174 | BC104726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P107807 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P107807 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439179 | NM_147111.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439179 | AF121985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439179 | BC104726.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25382065 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127314 | - | - |
| Whole Mouse Genome G4122A | A_51_P107807 | - | - |
| Whole Mouse Genome G4122A | A_51_P439179 | NM_147111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439179 | AF121985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439179 | BC104726.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129039 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112683 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264262 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264271 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264274 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264265 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264280 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06452461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06264264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25382045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06430421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06264256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06264275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06452493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06264262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06257020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06264266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06430420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06498086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25382056 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566200 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566199 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 467114 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6380465 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380465 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600115 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600115 | - | - |
| mouse-6 v1.1 (Illumina) | scl30933.2.1_243-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380465 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30933.2.1_243-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380465 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600115 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600115 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30933.2.1_243-S | - | - |

Olfr586
Genomic Map