Assay Details
| Gene Symbol: Olfr575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147114.2 | NP_667325.1 | 1 | 248 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961175 | NM_147114.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955378 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121457 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126719 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152628 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961175 | NM_147114.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381444 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121457 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152628 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126719 | - | - |
| Whole Mouse Genome G4122A | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272267 | NM_147114.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136275 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126719 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121457 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152628 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06566184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05908077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05998396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06090172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06374499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05916844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05981196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05916845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381437 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566191 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566190 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032678.mm.1 | NM_147114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032678.mm.1 | NM_147114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032678.mm.1 | TC0700003874.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 472203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4590402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4590402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712410 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2712410 | - | - |
| mouse-6 v1.1 (Illumina) | scl8888.1.1_269-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl8888.1.1_269-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2791541 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2791541 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl8888.1.1_269-S | - | - |

Olfr575
Genomic Map