Assay Details
Gene Symbol: Ly6g5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148947.1 | NP_683749.1 | 1 - 2 | 120 | 65 | |
GenBank mRNA | AJ315552.1 | - | 1 - 2 | 120 | 65 | |
BC145966.1 | - | 1 - 2 | 141 | 40131269 | 65 | |
BC145968.1 | - | 1 - 2 | 141 | 40131271 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
