Assay Details
| Gene Symbol: Ager | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271422.1 | NP_001258351.1 | 5 - 6 | 546 | 69 | |
| NM_001271423.1 | NP_001258352.1 | 5 - 6 | 519 | 69 | ||
| NM_001271424.1 | NP_001258353.1 | 4 - 5 | 641 | 69 | ||
| NM_007425.3 | NP_031451.2 | 5 - 6 | 546 | 69 | ||
| NR_073173.1 | 5 - 6 | 546 | 69 | |||
| NR_073175.1 | 4 - 5 | 741 | 69 | |||
| NR_073176.1 | 4 - 5 | 741 | 69 | |||
| GenBank mRNA | AB207883.1 | - | 5 - 6 | 506 | 69 | |
| BC061182.1 | - | 5 - 6 | 534 | 30300427 | 69 | |
| EU520325.1 | - | 5 - 6 | 502 | 69 | ||
| EU570240.1 | - | 5 - 6 | 502 | 69 | ||
| EU570241.1 | - | 4 - 5 | 697 | 69 | ||
| EU570242.1 | - | 5 - 6 | 502 | 69 | ||
| EU570243.1 | - | 5 - 6 | 502 | 69 | ||
| EU570244.1 | - | 4 - 5 | 697 | 69 | ||
| EU570245.1 | - | 4 - 5 | 697 | 69 | ||
| EU570246.1 | - | 4 - 5 | 659 | 69 | ||
| EU570247.1 | - | 4 - 5 | 697 | 69 | ||
| EU906862.1 | - | 5 - 6 | 502 | 69 | ||
| EU906863.1 | - | 5 - 6 | 475 | 69 | ||
| EU906864.1 | - | 4 - 5 | 697 | 69 | ||
| GU164727.1 | - | 4 - 5 | 616 | 69 | ||
| GU164728.1 | - | 5 - 6 | 521 | 69 | ||
| GU164729.1 | - | 5 - 6 | 521 | 69 | ||
| GU164730.1 | - | 5 - 6 | 521 | 69 | ||
| GU164731.1 | - | 5 - 6 | 521 | 69 | ||
| GU164732.1 | - | 5 - 6 | 521 | 69 | ||
| KC692918.1 | - | 5 - 6 | 502 | 69 | ||
| L33412.1 | - | 5 - 6 | 507 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ager
Genomic Map