Assay Details
| Gene Symbol: Serpina3m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009253.2 | NP_033279.2 | 63 | 74 | ||
| GenBank mRNA | AI286407.1 | - | 63 | 74 | ||
| BC011158.1 | - | 65 | 4194074 | 74 | ||
| BC053337.1 | - | 41 | 4208766 | 74 | ||
| X55148.1 | - | 35 | 74 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC011158.1,BC053337.1,NM_009253.2,X55148.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161410 | X55148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161410 | X69832.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161410 | NM_009253.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161410 | BC011158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161410 | BC053337.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632551 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171354 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114233 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161410 | X55148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161410 | X69832.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161410 | NM_009253.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161410 | BC011158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161410 | BC053337.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358012 | X55148.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702942 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07734453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02595137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114233 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07734436 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02595163 | - | - |
| Whole Mouse Genome G4122A | A_51_P358012 | X55148.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113518 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114233 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02595165 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171354 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09886645 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09886653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09886646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28703012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10230934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10127161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09886647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10127160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28703001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10020417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10131054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398073 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421921_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398069 | - | - |
| Mouse Genome 430 2.0 Array | 1421921_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421921_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20717_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20717_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007479.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007479.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007479.mm.1 | TC1200001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007483.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007483.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007483.mm.1 | TC1200001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007480.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007480.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007480.mm.1 | TC1200001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007482.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007482.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007482.mm.1 | TC1200001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007478.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007478.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007478.mm.1 | TC1200001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007477.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007477.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007477.mm.1 | TC1200001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007481.mm.1 | NM_009253.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007481.mm.1 | NM_009253.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007481.mm.1 | TC1200001044.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421921_PM_at | - | - |
| Mouse Expression Set 430 | 1421921_at | - | - |
| Murine Genome U74v2 Set | 102816_PM_at | - | - |
| Murine Genome U74v2 Set | 102816_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421921_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421921_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421921_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421921_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 545993 | - | - |
| ABI Mouse Genome Survey Microarray | 676305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 130253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 130253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000130253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2728466 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2728466 | - | - |
| mouse-6 v1.1 (Illumina) | scl020717.5_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 130253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020717.5_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2728473 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2728473 | - | - |

Serpina3m
Genomic Map