| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100008263.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008263.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008263.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008263.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008263.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008258.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008258.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008258.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008258.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008258.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008261.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008261.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008261.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008261.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008261.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008252.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008252.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008252.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008252.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008252.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008264.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008264.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008264.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008264.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008264.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008265.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008265.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008265.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008265.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008265.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008249.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008249.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008249.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008249.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008249.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008254.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008254.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008254.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008254.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008254.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008260.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008260.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008260.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008260.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008260.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008253.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008253.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008253.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008253.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008253.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008256.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008256.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008256.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008256.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008256.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008247.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008247.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008247.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008247.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008247.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008259.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008259.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008259.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008259.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008259.mm.1 |
TC1100000942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008250.mm.1 |
NM_001204252.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008250.mm.1 |
NM_001204252.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008250.mm.1 |
NM_010796.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008250.mm.1 |
NM_010796.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008250.mm.1 |
TC1100000942.mm.1 |
MTA TC |