Assay Details
| Gene Symbol: Tlr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016928.2 | NP_058624.2 | 4 | 1683 | 74 | |
| GenBank mRNA | AF186107.1 | - | 1 | 2352 | 74 | |
| AK036842.1 | - | 4 | 1685 | 74 | ||
| BC125240.1 | - | 2 | 1681 | 40043826 | 74 | |
| BC125247.1 | - | 2 | 1681 | 40055024 | 74 | |
| BC125261.1 | - | 2 | 1681 | 40048274 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cytokine production
immune system process
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
signal transduction
positive regulation of interleukin-8 production
positive regulation of toll-like receptor signaling pathway
toll-like receptor 5 signaling pathway
defense response to bacterium
innate immune response
positive regulation of nitric oxide biosynthetic process
regulation of cytokine secretion
transmembrane signaling receptor activity
interleukin-1 receptor binding
protein binding
positive regulation of cytokine production
immune system process
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
signal transduction
positive regulation of interleukin-8 production
positive regulation of toll-like receptor signaling pathway
toll-like receptor 5 signaling pathway
defense response to bacterium
innate immune response
positive regulation of nitric oxide biosynthetic process
regulation of cytokine secretion
transmembrane signaling receptor activity
interleukin-1 receptor binding
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131428 | BC125247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131428 | BC125261.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131428 | BC125240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131428 | NM_016928.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766762 | AK009705.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161723 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115910 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170125 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131428 | BC125247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131428 | BC125261.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131428 | BC125240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131428 | NM_016928.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115910 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170125 | - | - |
| Whole Mouse Genome G4122A | A_51_P461123 | AF186107.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111367 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126260 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170125 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20890008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20889970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00296755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00421387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00136069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00509123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00462221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00535473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00531995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00421379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00416606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00960862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00960853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00109228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00136076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00925198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20890101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20889995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00582407 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608697 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450242_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608697 | - | - |
| Mouse Genome 430 2.0 Array | 1450242_at | - | - |
| Mouse Genome 430 2.0 Array | 1450242_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100014643.mm.1 | TC0100001754.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014640.mm.1 | NM_016928.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014640.mm.1 | TC0100001754.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014641.mm.1 | NM_016928.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014641.mm.1 | TC0100001754.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014642.mm.1 | NM_016928.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014642.mm.1 | TC0100001754.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100014645.mm.1 | NM_016928.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100014645.mm.1 | TC0100001754.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450242_PM_at | - | - |
| Mouse Expression Set 430 | 1450242_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450242_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450242_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450242_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450242_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 639127 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1780131 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001780131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2440602 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2440602 | - | - |
| mouse-6 v1.1 (Illumina) | scl17112.5.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1780131 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17112.5.1_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780131 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2440602 | - | - |

Tlr5
Genomic Map