Assay Details
| Gene Symbol: Poteg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026256.2 | NP_080532.1 | 5 - 6 | 922 | 115 | |
| NM_027618.1 | NP_081894.1 | 5 - 6 | 922 | 115 | ||
| GenBank mRNA | AK015012.1 | - | 5 - 6 | 923 | 115 | |
| AK015165.1 | - | 5 - 6 | 923 | 115 | ||
| AK015473.1 | - | 5 - 6 | 923 | 115 | ||
| DQ298444.1 | - | 5 - 6 | 923 | 115 | ||
| DQ517440.1 | - | 5 - 6 | 923 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Poteg
Genomic Map