Assay Details
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163725.1 | NP_001157197.1 | 5 - 6 | 775 | 69 | |
NM_026314.3 | NP_080590.3 | 7 - 8 | 1135 | 69 | ||
GenBank mRNA | AK005832.1 | - | 4 - 5 | 585 | 69 | |
AK015345.1 | - | 3 - 4 | 314 | 69 | ||
AY428507.1 | - | 7 - 8 | 1135 | 69 | ||
AY464142.1 | - | 5 - 6 | 775 | 69 | ||
BC026462.1 | - | 7 - 8 | 1092 | 4483015 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
