Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 6 - 7 | 717 | 71 | |
| GenBank mRNA | AK011156.1 | - | 6 - 7 | 663 | 71 | |
| AK038318.1 | - | 6 - 7 | 695 | 71 | ||
| AK122320.1 | - | 5 - 6 | 723 | 71 | ||
| AK161181.1 | - | 6 - 7 | 666 | 71 | ||
| AK163765.1 | - | 5 - 6 | 705 | 71 | ||
| AK168388.1 | - | 6 - 7 | 704 | 71 | ||
| BC032177.1 | - | 4 - 5 | 944 | 71 | ||
| BC040393.1 | - | 6 - 7 | 641 | 71 | ||
| BC049979.1 | - | 6 - 7 | 693 | 71 | ||
| BC056942.1 | - | 6 - 7 | 644 | 6827177 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map