Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 13 - 14 | 1241 | 109 | |
| GenBank mRNA | AK038318.1 | - | 13 - 14 | 1219 | 109 | |
| AK043764.1 | - | 2 - 3 | 86 | 109 | ||
| AK122320.1 | - | 12 - 13 | 1247 | 109 | ||
| AK145716.1 | - | 13 - 14 | 1242 | 109 | ||
| AK161181.1 | - | 13 - 14 | 1191 | 109 | ||
| AK163765.1 | - | 12 - 13 | 1229 | 109 | ||
| AK168388.1 | - | 13 - 14 | 1228 | 109 | ||
| AK195523.1 | - | 82 | 109 | |||
| BC056942.1 | - | 13 - 14 | 1168 | 6827177 | 109 | |
More Information
Set Membership: |
|

Fam21
Genomic Map