Assay Details
| Gene Symbol: Agpat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026792.3 | NP_081068.1 | 4 - 5 | 773 | 101 | |
| GenBank mRNA | AK003649.1 | - | 4 - 5 | 517 | 101 | |
| AK082137.1 | - | 4 - 5 | 548 | 101 | ||
| AK089885.1 | - | 4 - 5 | 576 | 101 | ||
| AK152709.1 | - | 4 - 5 | 516 | 101 | ||
| AK152889.1 | - | 4 - 5 | 520 | 101 | ||
| AK155378.1 | - | 5 - 6 | 722 | 101 | ||
| AK170093.1 | - | 4 - 5 | 517 | 101 | ||
| AY161042.1 | - | 4 - 5 | 750 | 101 | ||
| BC062903.1 | - | 4 - 5 | 773 | 101 | ||
More Information
Set Membership: |
|

Agpat5
Genomic Map