Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100017251.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017251.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017251.mm.1 |
TC1100001832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017250.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017250.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017250.mm.1 |
TC1100001832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017243.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017243.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017243.mm.1 |
TC1100001832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017245.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017245.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017245.mm.1 |
TC1100001832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017252.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017252.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017252.mm.1 |
TC1100001832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017247.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017247.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017247.mm.1 |
TC1100001832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017248.mm.1 |
NM_028710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017248.mm.1 |
NM_028710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017248.mm.1 |
TC1100001832.mm.1 |
MTA TC |