Assay Details
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028710.3 | NP_082986.3 | 9 - 10 | 1766 | 79 | |
GenBank mRNA | AK018132.2 | - | 6 - 7 | 1183 | 79 | |
AK158726.1 | - | 9 - 10 | 1767 | 79 | ||
AK173082.1 | - | 6 - 7 | 865 | 79 | ||
AK186315.1 | - | 1 - 2 | 46 | 79 | ||
AK192114.1 | - | 1 - 2 | 172 | 79 | ||
BC022158.1 | - | 5 - 6 | 596 | 79 | ||
BC084731.1 | - | 6 - 7 | 1456 | 6813261 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
