Assay Details
| Gene Symbol: Eaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028932.4 | NP_083208.1 | 5 - 6 | 951 | 68 | |
| GenBank mRNA | AK016628.1 | - | 1632 | 68 | ||
| AK036661.1 | - | 5 - 6 | 923 | 68 | ||
| AK044181.1 | - | 5 - 6 | 952 | 68 | ||
| AK044737.1 | - | 5 - 6 | 888 | 68 | ||
| BC079658.1 | - | 5 - 6 | 928 | 6853920 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29512329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29512328 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29512331 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253080 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57518 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154460 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102146 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29512347 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102146 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08073091 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08073118 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154460 | - | - |
| Whole Mouse Genome G4122A | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | NM_028932.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | AK036661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | AK044181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | AK044737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419959 | BC079658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83386 | NM_028932.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83386 | AK016628.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83386 | AK036661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83386 | AK044181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83386 | AK044737.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123785 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154460 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11648923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11648915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11648926 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29512328 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29512331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29512329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29512355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11648926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11584385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11643179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11328582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11648915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29512354 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10413801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413802 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413796 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433555_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431065_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10413795 | - | - |
| Mouse Genome 430 2.0 Array | 1433555_at | - | - |
| Mouse Genome 430 2.0 Array | 1433555_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431065_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431065_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74427_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74427_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400002864.mm.1 | NM_028932.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002864.mm.1 | NM_028932.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002864.mm.1 | TC1400000329.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400002865.mm.1 | NM_028932.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002865.mm.1 | NM_028932.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002865.mm.1 | TC1400000329.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431065_at | - | - |
| Mouse Expression Set 430 | 1433555_PM_at | - | - |
| Mouse Expression Set 430 | 1433555_at | - | - |
| Mouse Expression Set 430 | 1431065_PM_at | - | - |
| Murine Genome U74v2 Set | 169274_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170089_PM_at | - | - |
| Murine Genome U74v2 Set | 170089_at | - | - |
| Murine Genome U74v2 Set | 169274_r_at | - | - |
| Murine Genome U74v2 Set | 168852_PM_at | - | - |
| Murine Genome U74v2 Set | 168852_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433555_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433555_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431065_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431065_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 322686 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6550619 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5420039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5420039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6550619 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550619 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223742 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232473 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232473 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223742 | - | - |
| mouse-6 v1.1 (Illumina) | scl000417.1_28-S | - | - |
| mouse-6 v1.1 (Illumina) | scl46488.7_372-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550619 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000417.1_28-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46488.7_372-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550619 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232473 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223742 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232473 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223742 | - | - |

Eaf1
Genomic Map