Assay Details
Gene Symbol: Cecr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033567.1 | NP_291045.1 | 1 | 4673 | 66 | |
GenBank mRNA | AF277399.1 | - | 1 | 4673 | 66 | |
AK140695.1 | - | 2 | 3743 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798779 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798785 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798775 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798778 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798783 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798776 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798782 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798784 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798781 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798787 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798780 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798786 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202971 | AK140695.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202971 | AK140695.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592882 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697424 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132081 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115906 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140026 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114664 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202971 | AK140695.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202971 | AF277399.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202971 | AK140695.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202971 | AK140695.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202971 | AK140695.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798793 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798813 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798779 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798801 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798806 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798804 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798777 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798796 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798788 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798797 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24798809 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140026 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115906 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114664 | - | - |
Whole Mouse Genome G4122A | A_51_P202971 | AK140695.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P202971 | AK140695.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P202971 | NM_033567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P202971 | AF277399.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111442 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140026 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114664 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115906 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05237847 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05237842 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05237841 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05237850 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798783 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798784 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798776 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798778 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798775 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798787 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798781 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798782 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798786 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798780 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798785 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24798779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24798793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05237847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P04998397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05576700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05526836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05526835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05752918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05110640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05575997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05563898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05454719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05563897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05563893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05441105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05752922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05118402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24798810 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10547509 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547510 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547511 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547512 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419224_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10547508 | - | - |
Mouse Genome 430 2.0 Array | 1419224_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419224_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94047_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94047_PM_at | - | - |
Mouse Expression Set 430 | 1419224_at | - | - |
Mouse Expression Set 430 | 1419224_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419224_at | - | - |
Mouse Genome 430A 2.0 Array | 1419224_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419224_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419224_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 797305 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 540672 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 540672 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000540672 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248101 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248101 | - | - |
mouse-6 v1.1 (Illumina) | scl28460.1.1_159-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 540672 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl28460.1.1_159-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 540672 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1248101 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248101 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28460.1.1_159-S | - | - |
