Assay Details
Gene Symbol: Imp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133976.2 | NP_598737.1 | 1 | 449 | 62 | |
GenBank mRNA | AK004456.1 | - | 1 | 476 | 62 | |
BC009145.1 | - | 1 | 452 | 2631069 | 62 |
Gene Symbol: 4933427G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030236.1 | - | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124497 | NM_133976.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124497 | AK004456.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124497 | AK209259.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124497 | BC009145.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124497 | NM_133976.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124497 | AK004456.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124497 | AK209259.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124497 | BC009145.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P284346 | NM_133976.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P284346 | AK004456.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P284346 | BC009145.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539451 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26539454 | - | - |
Whole Mouse Genome G4122A | A_52_P284346 | NM_133976.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P284346 | AK004456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P284346 | BC009145.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124497 | NM_133976.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124497 | AK004456.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124497 | AK209259.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124497 | BC009145.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26539408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08098443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08156596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07770485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539459 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07802664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26539430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08189251 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10585600 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416210_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10585599 | - | - |
Mouse Genome 430 2.0 Array | 1416210_at | - | - |
Mouse Genome 430 2.0 Array | 1416210_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102462_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102462_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900004984.mm.1 | NM_133976.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900004984.mm.1 | NM_133976.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900004984.mm.1 | TC0900000689.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416210_PM_at | - | - |
Mouse Expression Set 430 | 1416210_at | - | - |
Murine Genome U74v2 Set | 92817_at | - | - |
Murine Genome U74v2 Set | 92817_PM_at | - | - |
Murine Genome U74v2 Set | 171039_PM_at | - | - |
Murine Genome U74v2 Set | 171039_at | - | - |
Mouse Genome 430A 2.0 Array | 1416210_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416210_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416210_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416210_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 485001 | - | - |
ABI Mouse Genome Survey Microarray | 893995 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2900347 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2900347 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105860731 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002900347 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243632 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231341 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243632 | - | - |
mouse-6 v1.1 (Illumina) | GI_19527195-S | - | - |
mouse-6 v1.1 (Illumina) | scl0102462.1_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105860731 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2900347 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0102462.1_25-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105860731 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900347 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2593208 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2593208 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0102462.1_25-S | - | - |
