Assay Details
Gene Symbol: Rpp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133982.1 | NP_598743.1 | 1 | 453 | 105 | |
GenBank mRNA | AK082822.1 | - | 1 | 523 | 105 | |
AK140177.1 | - | 1 | 106 | |||
BC016085.1 | - | 1 | 453 | 4501535 | 105 | |
BC083064.1 | - | 1 | 513 | 5367203 | 105 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
